Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dimethylaminobenzoic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Plastic bottle
619-84-1
C9H11NO2
165.19
MFCD00002537
YDIYEOMDOWUDTJ-UHFFFAOYSA-N
4-dimethylamino benzoic acid,benzoic acid, 4-dimethylamino,benzoic acid, p-dimethylamino,p-dimethylaminobenzoic acid,n,n-dimethyl-p-aminobenzoic acid,n,n-dimethyl-4-aminobenzoic acid,p-n,n-dimethylamino benzoic acid,p-dimethylamino benzoic acid,unii-d1ma908ev0,4-n,n-dimethylamino benzoic acid
12092
4-(dimethylamino)benzoic acid
CN(C)C1=CC=C(C=C1)C(=O)O
619-84-1
C9H11NO2
165.19
MFCD00002537
YDIYEOMDOWUDTJ-UHFFFAOYSA-N
4-dimethylamino benzoic acid,benzoic acid, 4-dimethylamino,benzoic acid, p-dimethylamino,p-dimethylaminobenzoic acid,n,n-dimethyl-p-aminobenzoic acid,n,n-dimethyl-4-aminobenzoic acid,p-n,n-dimethylamino benzoic acid,p-dimethylamino benzoic acid,unii-d1ma908ev0,4-n,n-dimethylamino benzoic acid
12092
4-(dimethylamino)benzoic acid
CN(C)C1=CC=C(C=C1)C(=O)O
Flash Point 170°C
Color Beige-White to Gray-Green
Infrared Spectrum Authentic
Melting Point 238°C to 242°C
Density 170g/mL
Packaging Plastic bottle
Quantity 500g
Water 0.5% max. (K.F.)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3)2NC6H4CO2H
Beilstein 14, 426
Merck Index 15, 3254
Solubility Information Solubility in water: .. Other solubilities: soluble in hcl and koh solutions,soluble in alcohol,sparingly soluble in ether,practically insoluble in acetic acid
Formula Weight 165.19
Physical Form Crystalline Powder
Percent Purity ≥97.5% (on dry substance)
Chemical Name or Material 4-Dimethylaminobenzoic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning