Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dimethylaminobutyric acid hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7023251
Click to view available options
Quantity:
5 g
25 g
69954-66-1
C6H14ClNO2
167.633
MFCD00012613
RDTALXUBMCLWBB-UHFFFAOYSA-N
4-dimethylamino butanoic acid hydrochloride, 4-dimethylaminobutyric acid hydrochloride, 4-dimethylamino butyric acid hydrochloride, butanoic acid, 4-dimethylamino-, hydrochloride, 4-dimethylamino butanoic acid hcl, 4-dimethylamino butyric acid hcl salt, 4-dimethylamino-butyric acid hydrochloride, 4-dimethylamino-butyric acid hydrochloride salt
12691640
4-(dimethylamino)butanoic acid;hydrochloride
CN(C)CCCC(=O)O.Cl
69954-66-1
C6H14ClNO2
167.633
MFCD00012613
RDTALXUBMCLWBB-UHFFFAOYSA-N
4-dimethylamino butanoic acid hydrochloride, 4-dimethylaminobutyric acid hydrochloride, 4-dimethylamino butyric acid hydrochloride, butanoic acid, 4-dimethylamino-, hydrochloride, 4-dimethylamino butanoic acid hcl, 4-dimethylamino butyric acid hcl salt, 4-dimethylamino-butyric acid hydrochloride, 4-dimethylamino-butyric acid hydrochloride salt
12691640
4-(dimethylamino)butanoic acid;hydrochloride
CN(C)CCCC(=O)O.Cl

4-Dimethylaminobutyric acid hydrochloride is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Dimethylaminobutyric acid hydrochloride is used as pharmaceutical intermediates.

Notes
Hygroscopic. Protect from humidity and water. Store under dry inert gas. Incompatible with water and oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 153°C to 155°C
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 167.64
Percent Purity 98%
Chemical Name or Material 4-Dimethylaminobutyric acid hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit