Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dimethylaminocinnamaldehyde, 98+%, Affymetrix/USB™

Click to view available options
Quantity:
25g
100g
Packaging:
Plastic bottle
6203-18-5
C11H13NO
175.231
MFCD00007002
RUKJCCIJLIMGEP-ONEGZZNKSA-N
4-dimethylamino cinnamaldehyde,4-dimethylaminocinnamaldehyde,p-dimethylaminocinnamaldehyde,3-4-dimethylamino phenyl acrylaldehyde,4-dimethylcinnamaldehyde,p-dimethylaminocinnamic aldehyde,p-dimethylamino cinnamaldehyde,cinnamaldehyde, p-dimethylamino,unii-9rsi7wz9f0,dmaca reagent
5284506
(E)-3-[4-(dimethylamino)phenyl]prop-2-enal
CN(C)C1=CC=C(C=C1)C=CC=O
6203-18-5
C11H13NO
175.231
MFCD00007002
RUKJCCIJLIMGEP-ONEGZZNKSA-N
4-dimethylamino cinnamaldehyde,4-dimethylaminocinnamaldehyde,p-dimethylaminocinnamaldehyde,3-4-dimethylamino phenyl acrylaldehyde,4-dimethylcinnamaldehyde,p-dimethylaminocinnamic aldehyde,p-dimethylamino cinnamaldehyde,cinnamaldehyde, p-dimethylamino,unii-9rsi7wz9f0,dmaca reagent
5284506
(E)-3-[4-(dimethylamino)phenyl]prop-2-enal
CN(C)C1=CC=C(C=C1)C=CC=O
Quantity 25g
Melting Point 137°C to 144°C
Packaging Plastic bottle
Linear Formula (CH3)2NC6H4CH:CHCHO
Formula Weight 175.23
Physical Form Crystalline Powder
Percent Purity ≥98%
Chemical Name or Material 4-Dimethylaminocinnamaldehyde