Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dimethylaminopyridine (Peptide Synthesis), Fisher BioReagents

Click to view available options
Quantity:
100g
Packaging:
Glass Bottle
1122-58-3
C7H10N2
122.171
VHYFNPMBLIVWCW-UHFFFAOYSA-N
4-dimethylaminopyridine, dmap, 4-dimethylamino pyridine, 4-pyridinamine, n,n-dimethyl, n,n-dimethyl-4-pyridinamine, p-dimethylaminopyridine, pyridine, 4-dimethylamino, ccris 6176, dimethylpyridin-4-ylamine
14284
N,N-dimethylpyridin-4-amine
CN(C)C1=CC=NC=C1
1122-58-3
C7H10N2
122.171
VHYFNPMBLIVWCW-UHFFFAOYSA-N
4-dimethylaminopyridine, dmap, 4-dimethylamino pyridine, 4-pyridinamine, n,n-dimethyl, n,n-dimethyl-4-pyridinamine, p-dimethylaminopyridine, pyridine, 4-dimethylamino, ccris 6176, dimethylpyridin-4-ylamine
14284
N,N-dimethylpyridin-4-amine
CN(C)C1=CC=NC=C1
4-Dimethylaminopyridine is a catalyst for several acylation reactions, especially in oligonucleotide synthesis.
Alpha Vector Dimethylaminopyridine
Melting Point 110°C
Identification Pass Test
Packaging Glass Bottle
Boiling Point 162°C
Color White
pH 9.7
Quantity 100g
Assay Percent Range ≥99 %
Merck Index 15, 3433
Formula Weight 122.17
Physical Form Solid
Percent Purity ≥99%
Grade Peptide Synthesis
Chemical Name or Material 4-Dimethylaminopyridine
Show More Show Less

Recommended Storage : RT