Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Diphenylamino)benzaldehyde, 98%

Click to view available options
Quantity:
1g
5g
25g
4181-05-9
C19H15NO
273.34
MFCD00145131
UESSERYYFWCTBU-UHFFFAOYSA-N
4-diphenylamino benzaldehyde,4-n,n-diphenylamino benzaldehyde,4-diphenylaminobenzaldehyde,4-diphenylamino-benzaldehyde,benzaldehyde, 4-diphenylamino,4-formyltriphenylamine,p-formyltriphenylamine,4-n-phenylanilino benzaldehyde,p-diphenylaminobenzaldehyde,4-n,n-diphenylamino-benzaldehyde
77846
4-(N-phenylanilino)benzaldehyde
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C=O
4181-05-9
C19H15NO
273.34
MFCD00145131
UESSERYYFWCTBU-UHFFFAOYSA-N
4-diphenylamino benzaldehyde,4-n,n-diphenylamino benzaldehyde,4-diphenylaminobenzaldehyde,4-diphenylamino-benzaldehyde,benzaldehyde, 4-diphenylamino,4-formyltriphenylamine,p-formyltriphenylamine,4-n-phenylanilino benzaldehyde,p-diphenylaminobenzaldehyde,4-n,n-diphenylamino-benzaldehyde
77846
4-(N-phenylanilino)benzaldehyde
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C=O
Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Melting Point 129°C to 133°C (literature)
Color Yellow
Odor Odorless
Quantity 5g
Beilstein 2732795
Formula Weight 273.33
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 4-(Diphenylamino)benzaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

storageNote1 : 2° to 8°C