Alfa Aesar™ 4-Ethoxy-1,1,1-trifluoro-3-buten-2-one, 94%, stab.

Manufacturer: Alfa Aesar™ H2697506

Quantity 5g

 View more versions of this product

Catalog No. AAH2697506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17129-06-5
    Molecular Formula C6H7F3O2
    Formula Weight 168.13
    MDL Number MFCD00192131
    UN Number UN3384
    Synonym ETFBO
    Chemical Name or Material 4-Ethoxy-1,1,1-trifluoro-3-buten-2-one, stabilized

    Specifications

    Formula Weight 168.13
    Molecular Formula C6H7F3O2
    UN Number UN3384
    Synonym ETFBO
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P280h-P273-P305+P351+P338-P304+P340-P309-P310
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Recommended Storage −30° to −10°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S.
    Flash Point 52°C (126°F)
    Odor Ether-like
    Boiling Point 159°C
    Melting Point −30°C
    Quantity 5g
    MDL Number MFCD00192131
    CAS 17129-06-5
    Health Hazard 2 GHS H Statement
    H330-H372-H226-H302-H315-H319-H317
    Fatal if inhaled.
    Causes damage to organs through prolonged or repeated exposure.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    Chemical Name or Material 4-Ethoxy-1,1,1-trifluoro-3-buten-2-one, stabilized
    Assay 94%

    GHS H Statement
    H330-H372-H226-H302-H315-H319-H317
    Fatal if inhaled.
    Causes damage to organs through prolonged or repeated exposure.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.

    −30° to −10°C

    GHS P Statement
    P280h-P273-P305+P351+P338-P304+P340-P309-P310
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit