Alfa Aesar™ 4-Ethoxy-2,3-difluorobenzoic acid, 98%

Manufacturer: Alfa Aesar™ H2608903

Quantity 1g

 View more versions of this product

Catalog No. AAH2608903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 124728-45-6
    Molecular Formula C9H8F2O3
    Formula Weight 202.15
    MDL Number MFCD02258849
    Synonym 2,3-Difluoro-4-ethoxybenzoic acid
    Chemical Name or Material 4-Ethoxy-2,3-difluorobenzoic acid

    Specifications

    Formula Weight 202.15
    Molecular Formula C9H8F2O3
    MDL Number MFCD02258849
    Synonym 2,3-Difluoro-4-ethoxybenzoic acid
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 200° to 203°C
    Quantity 1g
    CAS 124728-45-6
    Health Hazard 1 Warning
    Chemical Name or Material 4-Ethoxy-2,3-difluorobenzoic acid
    Assay 98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit