Alfa Aesar™ 4-Ethoxy-2,6-difluorobenzyl bromide, 97%

Manufacturer: Alfa Aesar™ H32084MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH32084MD

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C9H9BrF2O
    Formula Weight 251.07
    MDL Number MFCD09258697
    UN Number UN3261
    Chemical Name or Material 4-Ethoxy-2,6-difluorobenzyl bromide

    Specifications

    Formula Weight 251.07
    Molecular Formula C9H9BrF2O
    MDL Number MFCD09258697
    UN Number UN3261
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Quantity 250mg
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
    Melting Point 38° to 41°C
    Chemical Name or Material 4-Ethoxy-2,6-difluorobenzyl bromide
    Assay 97%

    Description

    Applications
    4-Ethoxy-2,6-difluorobenzyl bromide, is used as an intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit