Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxy-2-nitroaniline, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
616-86-4
C8H10N2O3
182.179
MFCD00007153
ISFYBUAVOZFROB-UHFFFAOYSA-N
2-nitro-p-phenetidine,benzenamine, 4-ethoxy-2-nitro,4-amino-3-nitrophenetole,p-phenetidine, 2-nitro,unii-k7399mo0i4,2-nitro-4-ethoxyaniline,4-ethyloxy-2-nitroaniline,4-ethoxy-2-nitrophenyl amine,4-ethoxy-2-nitrophenylamine,acmc-20amjk
69229
4-ethoxy-2-nitroaniline
CCOC1=CC(=C(C=C1)N)[N+](=O)[O-]
616-86-4
C8H10N2O3
182.179
MFCD00007153
ISFYBUAVOZFROB-UHFFFAOYSA-N
2-nitro-p-phenetidine,benzenamine, 4-ethoxy-2-nitro,4-amino-3-nitrophenetole,p-phenetidine, 2-nitro,unii-k7399mo0i4,2-nitro-4-ethoxyaniline,4-ethyloxy-2-nitroaniline,4-ethoxy-2-nitrophenyl amine,4-ethoxy-2-nitrophenylamine,acmc-20amjk
69229
4-ethoxy-2-nitroaniline
CCOC1=CC(=C(C=C1)N)[N+](=O)[O-]
Infrared Spectrum Authentic
Color Brown-Rust to Red
Melting Point 110°C to 114°C
Quantity 1g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C2H5OC6H3(NO2)NH2
Beilstein 13, 521
Formula Weight 182.18
Physical Form Crystals or Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 4-Ethoxy-2-nitroaniline, 97%
Show More Show Less