Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxy-3-methoxybenzaldehyde, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
Plastic bottle
120-25-2
C10H12O3
180.203
MFCD00016614
BERFDQAMXIBOHM-UHFFFAOYSA-N
4-ethoxy-3-anisaldehyde,4-ethoxy-m-anisaldehyde,benzaldehyde, 4-ethoxy-3-methoxy,ethyl-vanillin,asischem n42243,4-ethoxy-3-methoxy-benzaldehyde,ethylvanillin?,4-ethoxyvanillin,vanillin ethyl ether,pubchem2648
67116
4-ethoxy-3-methoxybenzaldehyde
CCOC1=C(C=C(C=C1)C=O)OC
120-25-2
C10H12O3
180.203
MFCD00016614
BERFDQAMXIBOHM-UHFFFAOYSA-N
4-ethoxy-3-anisaldehyde,4-ethoxy-m-anisaldehyde,benzaldehyde, 4-ethoxy-3-methoxy,ethyl-vanillin,asischem n42243,4-ethoxy-3-methoxy-benzaldehyde,ethylvanillin?,4-ethoxyvanillin,vanillin ethyl ether,pubchem2648
67116
4-ethoxy-3-methoxybenzaldehyde
CCOC1=C(C=C(C=C1)C=O)OC
Color Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Flash Point 110°C
Melting Point 60.0°C to 65.0°C
Boiling Point 168.0°C
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula C2H5OC6H3(OCH3)CHO
Formula Weight 180.2
Physical Form Fine Crystalline Powder
Percent Purity 95%
Chemical Name or Material 4-Ethoxy-3-methoxybenzaldehyde, 95%
Show More Show Less

EINECSNumber : 204-382-1

TSCA : TSCA