Alfa Aesar™ 4-Ethoxy-3-methoxybenzaldehyde, 98+%

Manufacturer: Alfa Aesar™ A1089736

Quantity 500g

 View more versions of this product

Catalog No. AAA1089736

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 120-25-2
    Molecular Formula C10H12O3
    Formula Weight 180.2
    MDL Number MFCD00016614
    Beilstein 391169
    Synonym 4-Ethoxy-m-anisaldehyde
    Chemical Name or Material 4-Ethoxy-3-methoxybenzaldehyde

    Specifications

    Formula Weight 180.2
    Quantity 500g
    Molecular Formula C10H12O3
    MDL Number MFCD00016614
    CAS 120-25-2
    Beilstein 391169
    Melting Point 59° to 63°C
    TSCA TSCA
    Synonym 4-Ethoxy-m-anisaldehyde
    Chemical Name or Material 4-Ethoxy-3-methoxybenzaldehyde
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit