Alfa Aesar™ 4-Ethoxy-3-(trifluoromethyl)aniline, 97%

Manufacturer: Alfa Aesar™ H3443903

Quantity 1g

 View more versions of this product

Catalog No. AAH3443903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2713-74-8
    Molecular Formula C9H10F3NO
    Formula Weight 205.18
    MDL Number MFCD08056137
    UN Number UN2811
    Chemical Name or Material 4-Ethoxy-3-(trifluoromethyl)aniline

    Specifications

    Formula Weight 205.18
    Molecular Formula C9H10F3NO
    MDL Number MFCD08056137
    UN Number UN2811
    Health Hazard 2 GHS H Statement
    H331-H302-H312
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Quantity 1g
    CAS 2713-74-8
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material 4-Ethoxy-3-(trifluoromethyl)aniline
    Assay 97%

    GHS H Statement
    H331-H302-H312
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P261-P280-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit