Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxybenzaldehyde, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
10031-82-0
C9H10O2
150.18
MFCD00003388
JRHHJNMASOIRDS-UHFFFAOYSA-N
benzaldehyde, 4-ethoxy,p-ethoxybenzaldehyde,ethoxybenzaldehyde,benzaldehyde, p-ethoxy,homoanisaldehyde,4-ethyloxy benzaldehyde,unii-gow1h0f49a,fema no. 2413,4-ethoxy-benzaldehyde,ethoxybenzaldehyde, p
24834
4-ethoxybenzaldehyde
CCOC1=CC=C(C=C1)C=O
10031-82-0
C9H10O2
150.18
MFCD00003388
JRHHJNMASOIRDS-UHFFFAOYSA-N
benzaldehyde, 4-ethoxy,p-ethoxybenzaldehyde,ethoxybenzaldehyde,benzaldehyde, p-ethoxy,homoanisaldehyde,4-ethyloxy benzaldehyde,unii-gow1h0f49a,fema no. 2413,4-ethoxy-benzaldehyde,ethoxybenzaldehyde, p
24834
4-ethoxybenzaldehyde
CCOC1=CC=C(C=C1)C=O
Boiling Point 255°C
Density 1.0800g/mL
Melting Point 13°C to 14°C
Specific Gravity 1.08
Flash Point 75°C
Refractive Index 1.5574 to 1.5594
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
Color Yellow to Brown
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula C2H5OC6H4CHO
Beilstein 08,73
Formula Weight 150.18
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 4-Ethoxybenzaldehyde, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 233-093-3

RTECSNumber : CU6100000

TSCA : TSCA

Recommended Storage : this product darkens on storage