Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Ethoxybenzeneboronic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
22237-13-4
C8H11BO3
165.983
MFCD00674028
WRQNDLDUNQMTCL-UHFFFAOYSA-N
4-ethoxybenzeneboronic acid,4-ethoxyphenyl boronic acid,4-ethoxyphenyl boranediol,p-ethoxyphenylboronic acid,4-n-ethoxyphenyl boronic acid,boronic acid, 4-ethoxyphenyl,4-ethoxybenzeneboronicacid,4-ethoxyphenylboronicacid,4-ethoxyphenylboronic
2734351
(4-ethoxyphenyl)boronic acid
B(C1=CC=C(C=C1)OCC)(O)O
22237-13-4
C8H11BO3
165.983
MFCD00674028
WRQNDLDUNQMTCL-UHFFFAOYSA-N
4-ethoxybenzeneboronic acid,4-ethoxyphenyl boronic acid,4-ethoxyphenyl boranediol,p-ethoxyphenylboronic acid,4-n-ethoxyphenyl boronic acid,boronic acid, 4-ethoxyphenyl,4-ethoxybenzeneboronicacid,4-ethoxyphenylboronicacid,4-ethoxyphenylboronic
2734351
(4-ethoxyphenyl)boronic acid
B(C1=CC=C(C=C1)OCC)(O)O
Quantity 5g
Melting Point 165°C to 168°C
Beilstein 3530408
Formula Weight 165.99
Percent Purity 98%
Chemical Name or Material 4-Ethoxybenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning