Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Ethoxybenzyl alcohol, 98%

Manufacturer:  Alfa Aesar™ A1033109

Quantity 10g

 View more versions of this product

Catalog No. AAA1033109


  Check Availability
Add to cart

Specifications

Description

Applications
It is used in the identification of sulfation sites of metabolites and prediction of the compounds’ biological effects and in chemical investigation of Wedelia calendulacea.

Solubility
Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications

Chemical Identifiers

6214-44-4
152.19
4-Ethoxybenzyl alcohol
C9H12O2
MFCD00004655

Specifications

6214-44-4
152.19
272° to 274°C
10g
4-Ethoxybenzyl alcohol
C9H12O2
MFCD00004655
32° to 34°C
>110°C (230°F)
98%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit