Alfa Aesar™ 4-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, 97%

Manufacturer: Alfa Aesar™ H5583603

Quantity 1g

 View more versions of this product

Catalog No. AAH5583603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 195062-62-5
    Formula Weight 276.14
    MDL Number MFCD03453659
    Synonym 4-Ethoxycarbonylphenylboronic acid pinacol ester
    Chemical Name or Material 4-(Ethoxycarbonyl)benzeneboronic acid pinacol ester

    Specifications

    Formula Weight 276.14
    Synonym 4-Ethoxycarbonylphenylboronic acid pinacol ester
    Refractive Index 1.5000
    Physical Form Liquid
    MDL Number MFCD03453659
    CAS 195062-62-5
    Color Colorless
    Density 1.052g/mL
    Boiling Point 330°C
    Quantity 1g
    Chemical Name or Material 4-(Ethoxycarbonyl)benzeneboronic acid pinacol ester
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit