Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxycarbonylphenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
4334-88-7
C9H11BO4
193.99
MFCD02179441
ZLNFACCFYUFTLD-UHFFFAOYSA-N
4-ethoxycarbonylphenyl boronic acid,4-ethoxycarbonyl phenylboronic acid,4-ethoxycarbonyl phenyl boronic acid,4-ethoxycarbonyl benzeneboronic acid,4-ethoxycarbonylbenzeneboronic acid,ethyl 4-boronobenzoate,p-ethoxycarbonylphenylboronic acid,ethyl 4-dihydroxyboranyl benzoate,4-carbethoxyphenylboronic acid,benzoic acid, 4-borono-, 1-ethyl ester
2734350
(4-ethoxycarbonylphenyl)boronic acid
B(C1=CC=C(C=C1)C(=O)OCC)(O)O
4334-88-7
C9H11BO4
193.99
MFCD02179441
ZLNFACCFYUFTLD-UHFFFAOYSA-N
4-ethoxycarbonylphenyl boronic acid,4-ethoxycarbonyl phenylboronic acid,4-ethoxycarbonyl phenyl boronic acid,4-ethoxycarbonyl benzeneboronic acid,4-ethoxycarbonylbenzeneboronic acid,ethyl 4-boronobenzoate,p-ethoxycarbonylphenylboronic acid,ethyl 4-dihydroxyboranyl benzoate,4-carbethoxyphenylboronic acid,benzoic acid, 4-borono-, 1-ethyl ester
2734350
(4-ethoxycarbonylphenyl)boronic acid
B(C1=CC=C(C=C1)C(=O)OCC)(O)O
Melting Point 184°C to 189°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color White
Quantity 1g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 193.99
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 4-Ethoxycarbonylphenylboronic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning