Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxymethylene-2-phenyl-2-oxazolin-5-one, ≥90% (HPLC), MilliporeSigma™ Supelco™

$45.45 - $225.00

Specifications

Percent Purity ≥90% (HPLC)
Grade Analytical Standard (for drug analysis)
Linear Formula C12H11 NO3
Physical Form Neat
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-3246
View Documents
MilliporeSigma Supelco
E07531G
1 g
Each for $45.45
Add to cart
 
11-101-3248
View Documents
MilliporeSigma Supelco
E07535G
5 g
Each for $177.50
Add to cart
 
11-101-3247
View Documents
MilliporeSigma Supelco
E075310G
10 g
Each for $225.00
Add to cart
 
Description

Description

Pharmaceutical (small molecule)
Specifications

Specifications

≥90% (HPLC)
Analytical Standard (for drug analysis)
C12H11 NO3
Neat
Safety and Handling

Safety and Handling

RTECSNumber : RQ5786250

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit