Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxyphenylboronic acid, 97%, May contain varying amounts of anhydride, Maybridge

Click to view available options
Quantity:
1g
Packaging:
Amber Glass Bottle
22237-13-4
C8H11BO3
165.983
MFCD00674028
WRQNDLDUNQMTCL-UHFFFAOYSA-N
4-ethoxybenzeneboronic acid,4-ethoxyphenyl boronic acid,4-ethoxyphenyl boranediol,p-ethoxyphenylboronic acid,4-n-ethoxyphenyl boronic acid,boronic acid, 4-ethoxyphenyl,4-ethoxybenzeneboronicacid,4-ethoxyphenylboronicacid,4-ethoxyphenylboronic
2734351
(4-ethoxyphenyl)boronic acid
B(C1=CC=C(C=C1)OCC)(O)O
22237-13-4
C8H11BO3
165.983
MFCD00674028
WRQNDLDUNQMTCL-UHFFFAOYSA-N
4-ethoxybenzeneboronic acid,4-ethoxyphenyl boronic acid,4-ethoxyphenyl boranediol,p-ethoxyphenylboronic acid,4-n-ethoxyphenyl boronic acid,boronic acid, 4-ethoxyphenyl,4-ethoxybenzeneboronicacid,4-ethoxyphenylboronicacid,4-ethoxyphenylboronic
2734351
(4-ethoxyphenyl)boronic acid
B(C1=CC=C(C=C1)OCC)(O)O
Quantity 1g
Packaging Amber Glass Bottle
Infrared Spectrum Authentic
Color White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 165.98
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 4-Ethoxyphenylboronic acid
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.