Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxystyrene, 98%, stabilized, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
5459-40-5
C10H12O
148.205
MFCD00009097
OBRYRJYZWVLVLF-UHFFFAOYSA-N
4-ethoxystyrene,p-ethoxystyrene,p-vinylphenetole,benzene, 1-ethenyl-4-ethoxy,1-ethoxy-4-vinylbenzene,acmc-20amj9,1-ethoxy-4-vinylbenzene #,benzene,1-ethenyl-4-ethoxy,4-ethoxystyrene, technical grade
79570
1-ethenyl-4-ethoxybenzene
CCOC1=CC=C(C=C1)C=C
5459-40-5
C10H12O
148.205
MFCD00009097
OBRYRJYZWVLVLF-UHFFFAOYSA-N
4-ethoxystyrene,p-ethoxystyrene,p-vinylphenetole,benzene, 1-ethenyl-4-ethoxy,1-ethoxy-4-vinylbenzene,acmc-20amj9,1-ethoxy-4-vinylbenzene #,benzene,1-ethenyl-4-ethoxy,4-ethoxystyrene, technical grade
79570
1-ethenyl-4-ethoxybenzene
CCOC1=CC=C(C=C1)C=C
Stabilizer 0.2 to 0.5% t-butylcatechol
Color Colorless
Infrared Spectrum Authentic
Refractive Index 1.5490 to 1.5510 (20°C, 589nm)
Packaging Glass bottle
Density 0.9900g/mL
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula H3CCH2OC6H4CHCH2
Formula Weight 148.2
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 4-Ethoxystyrene, 98%
Show More Show Less