Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethyl-4-formylhexanenitrile, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2938-69-4
C9H15NO
153.22
MFCD00047654
AMUXIQHQXOKMTN-UHFFFAOYSA-N
4-cyano-2,2-diethylbutyraldehyde, acmc-1cbzp, 4-cyano-2,2-diethylbutanal, hexanenitrile,4-ethyl-4-formyl, hexanenitrile, 4-ethyl-4-formyl, 3-2-cyanoethyl-3-formylpentane, 2-beta-cyanoethyl-2-ethyl butyraldehyde
76248
4-ethyl-4-formylhexanenitrile
CCC(CC)(CCC#N)C=O
2938-69-4
C9H15NO
153.22
MFCD00047654
AMUXIQHQXOKMTN-UHFFFAOYSA-N
4-cyano-2,2-diethylbutyraldehyde, acmc-1cbzp, 4-cyano-2,2-diethylbutanal, hexanenitrile,4-ethyl-4-formyl, hexanenitrile, 4-ethyl-4-formyl, 3-2-cyanoethyl-3-formylpentane, 2-beta-cyanoethyl-2-ethyl butyraldehyde
76248
4-ethyl-4-formylhexanenitrile
CCC(CC)(CCC#N)C=O
Packaging Glass bottle
Infrared Spectrum Authentic
Specific Gravity 0.948
Boiling Point 94°C to 99°C (1.0mmHg)
Flash Point 96°C
Density 0.9480g/mL
Refractive Index 1.4505 to 1.4525
Color Colorless to Yellow
Quantity 25g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Linear Formula (C2H5)2C(CHO)CH2CH2CN
Formula Weight 153.22
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 4-Ethyl-4-formylhexanenitrile, 95%
Show More Show Less

STENCH

EINECSNumber : 220-923-4