Alfa Aesar™ 4-Ethyl-trans-beta-styrylboronic acid pinacol ester, 97%

Manufacturer: Alfa Aesar™ H3074403

Quantity 1g

 View more versions of this product

Catalog No. AAH3074403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 870717-91-2
    Molecular Formula C16H23BO2
    Formula Weight 258.17
    MDL Number MFCD08276837
    Synonym 2-[trans-2-(4-Ethylphenyl)vinyl]-4,4,5,5-tetramethyl[1,3,2]dioxaborolane
    Chemical Name or Material 4-Ethyl-trans-beta-styrylboronic acid pinacol ester

    Specifications

    Formula Weight 258.17
    Molecular Formula C16H23BO2
    MDL Number MFCD08276837
    CAS 870717-91-2
    Synonym 2-[trans-2-(4-Ethylphenyl)vinyl]-4,4,5,5-tetramethyl[1,3,2]dioxaborolane
    Melting Point 46° to 48°C
    Quantity 1g
    Flash Point >110°C (230°F)
    Chemical Name or Material 4-Ethyl-trans-beta-styrylboronic acid pinacol ester
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit