Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4'-Ethylacetophenone, 97%

Click to view available options
Quantity:
25g
100g
937-30-4
C10H12O
148.205
MFCD00009262
NODGRWCMFMEGJH-UHFFFAOYSA-N
4'-ethylacetophenone,1-4-ethylphenyl ethanone,p-ethylacetophenone,4-ethylacetophenone,ethanone, 1-4-ethylphenyl,p-acetylethylbenzene,acetophenone, 4'-ethyl,p-ethylphenyl methyl ketone,1-4-ethylphenyl ethan-1-one
13642
1-(4-ethylphenyl)ethanone
CCC1=CC=C(C=C1)C(=O)C
937-30-4
C10H12O
148.205
MFCD00009262
NODGRWCMFMEGJH-UHFFFAOYSA-N
4'-ethylacetophenone,1-4-ethylphenyl ethanone,p-ethylacetophenone,4-ethylacetophenone,ethanone, 1-4-ethylphenyl,p-acetylethylbenzene,acetophenone, 4'-ethyl,p-ethylphenyl methyl ketone,1-4-ethylphenyl ethan-1-one
13642
1-(4-ethylphenyl)ethanone
CCC1=CC=C(C=C1)C(=O)C
Applications
It can be used to produce 1,1′-p-phenylene-bis-ethanone. This reaction will need reagent air, cobalt naphthenate. The reaction temperature is 125°C.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Boiling Point 239°C
Quantity 100g
Flash Point 90°C (194°F)
Beilstein 1906029
Formula Weight 148.21
Percent Purity 97%
Chemical Name or Material 4'-Ethylacetophenone

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA