Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Ethylaminomethyl)pyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
50g
33403-97-3
C8H12N2
136.198
MFCD00023632
ZBAMQLFFVBPAOX-UHFFFAOYSA-N
4-ethylaminomethyl pyridine,n-pyridin-4-ylmethyl ethanamine,n-4-pyridylmethyl ethylamine,4-pyridinemethanamine, n-ethyl,unii-eye32x3w6x,n-ethyl-4-pyridinemethanamine,ethyl pyridin-4-ylmethyl amine,4-ethylamino methyl pyridine,n-ethyl-4-picolylamine,eye32x3w6x
96681
N-(pyridin-4-ylmethyl)ethanamine
CCNCC1=CC=NC=C1
33403-97-3
C8H12N2
136.198
MFCD00023632
ZBAMQLFFVBPAOX-UHFFFAOYSA-N
4-ethylaminomethyl pyridine,n-pyridin-4-ylmethyl ethanamine,n-4-pyridylmethyl ethylamine,4-pyridinemethanamine, n-ethyl,unii-eye32x3w6x,n-ethyl-4-pyridinemethanamine,ethyl pyridin-4-ylmethyl amine,4-ethylamino methyl pyridine,n-ethyl-4-picolylamine,eye32x3w6x
96681
N-(pyridin-4-ylmethyl)ethanamine
CCNCC1=CC=NC=C1
Color Colorless to Yellow
Refractive Index 1.5150 to 1.5170 (20°C, 589nm)
Density 0.98g/mL
Infrared Spectrum Authentic
Quantity 50g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 136.2
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 4-(Ethylaminomethyl)pyridine, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger