Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Ethylaniline, 98+%

Manufacturer:  Alfa Aesar™ B24652-22

Quantity 100g

 View more versions of this product

Catalog No. AAB2465222

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

589-16-2
121.18
UN2810
4-Ethylaniline
C8H11N
MFCD00007921
774319

Specifications

589-16-2
217° to 218°C
100g
Characteristic
MFCD00007921
774319
≥98%
−5°C
C8H11N
97°C (207°F)
121.18
UN2810
4-Ethylaniline

Safety and Handling

GHS H Statement
H301-H331-H373-H312
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.

GHS P Statement
P260-P261-P280-P301+P310-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit