Alfa Aesar™ 4-Ethylaniline, 98+%

Manufacturer: Alfa Aesar™ B24652-36

Quantity 500g

 View more versions of this product

Catalog No. AAB2465236

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 589-16-2
    Molecular Formula C8H11N
    Formula Weight 121.18
    MDL Number MFCD00007921
    UN Number UN2810
    Beilstein 774319
    Chemical Name or Material 4-Ethylaniline

    Specifications

    Formula Weight 121.18
    Quantity 500g
    Flash Point 97°C (207°F)
    Odor Characteristic
    MDL Number MFCD00007921
    CAS 589-16-2
    UN Number UN2810
    Beilstein 774319
    TSCA TSCA
    Melting Point −5°C
    Boiling Point 217° to 218°C
    Molecular Formula C8H11N
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H331-H373-H312
    Toxic if swallowed.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful in contact with skin.
    Health Hazard 3 GHS P Statement
    P260-P261-P280-P301+P310-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Chemical Name or Material 4-Ethylaniline
    Assay ≥98%

    GHS H Statement
    H301-H331-H373-H312
    Toxic if swallowed.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful in contact with skin.

    GHS P Statement
    P260-P261-P280-P301+P310-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit