Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethylaniline, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
589-16-2
C8H11N
121.183
MFCD00007921
HRXZRAXKKNUKRF-UHFFFAOYSA-N
p-ethylaniline,benzenamine, 4-ethyl,4-ethylphenylamine,aniline, 4-ethyl,p-ethylaminobenzene,1-amino-4-ethylbenzene,4-aminoethylbenzene,aniline, p-ethyl,unii-mhx72w1zqv,ccris 5060
11504
4-ethylaniline
CCC1=CC=C(C=C1)N
589-16-2
C8H11N
121.183
MFCD00007921
HRXZRAXKKNUKRF-UHFFFAOYSA-N
p-ethylaniline,benzenamine, 4-ethyl,4-ethylphenylamine,aniline, 4-ethyl,p-ethylaminobenzene,1-amino-4-ethylbenzene,4-aminoethylbenzene,aniline, p-ethyl,unii-mhx72w1zqv,ccris 5060
11504
4-ethylaniline
CCC1=CC=C(C=C1)N
Quantity 2.5kg
Infrared Spectrum Authentic
Density 0.97g/mL
Color Undesignated
Packaging Glass bottle
Refractive Index 1.5532 to 1.5552 (20°C, 589nm)
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula C2H5C6H4NH2
Beilstein 12, 1090
Formula Weight 121.18
Physical Form Liquid
Percent Purity ≥99.0%
Chemical Name or Material 4-Ethylaniline, 99+%
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-637-0

RTECSNumber : BX9900000

TSCA : TSCA