Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Ethylbenzeneboronic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
63139-21-9
C8H11BO2
149.984
MFCD00859377
RZCPLOMUUCFPQA-UHFFFAOYSA-N
4-ethylphenyl boronic acid,4-ethylbenzeneboronic acid,4-ethylphenyl boranediol,4-ethylphenylboronicacid,p-ethylphenylboronic acid,boronic acid, 4-ethylphenyl,pubchem7884,4-ethylbenzenboronic acid,acmc-1bik6
2734352
(4-ethylphenyl)boronic acid
B(C1=CC=C(C=C1)CC)(O)O
63139-21-9
C8H11BO2
149.984
MFCD00859377
RZCPLOMUUCFPQA-UHFFFAOYSA-N
4-ethylphenyl boronic acid,4-ethylbenzeneboronic acid,4-ethylphenyl boranediol,4-ethylphenylboronicacid,p-ethylphenylboronic acid,boronic acid, 4-ethylphenyl,pubchem7884,4-ethylbenzenboronic acid,acmc-1bik6
2734352
(4-ethylphenyl)boronic acid
B(C1=CC=C(C=C1)CC)(O)O
Melting Point 150°C to 154°C
Quantity 1g
Beilstein 3030638
Formula Weight 149.99
Percent Purity 97%
Chemical Name or Material 4-Ethylbenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning