Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Ethylbenzenesulfonyl chloride, 97%

Click to view available options
Quantity:
5g
25g
100g
16712-69-9
C8H9ClO2S
204.668
MFCD00041480
LACFLXDRFOQEFZ-UHFFFAOYSA-N
4-ethylbenzene-1-sulfonyl chloride,p-ethylbenzenesulfonyl chloride,4-ethylbenzenesulphonyl chloride,4-ethyl-benzenesulfonyl chloride,ethylbenzenesulfonylchloride,4-ethylbenzenesulfonylchloride,chloro 4-ethylphenyl sulfone,pubchem5133,acmc-209dvn,4-ethylphenylsulfonyl chloride
570959
4-ethylbenzenesulfonyl chloride
CCC1=CC=C(C=C1)S(=O)(=O)Cl
16712-69-9
C8H9ClO2S
204.668
MFCD00041480
LACFLXDRFOQEFZ-UHFFFAOYSA-N
4-ethylbenzene-1-sulfonyl chloride,p-ethylbenzenesulfonyl chloride,4-ethylbenzenesulphonyl chloride,4-ethyl-benzenesulfonyl chloride,ethylbenzenesulfonylchloride,4-ethylbenzenesulfonylchloride,chloro 4-ethylphenyl sulfone,pubchem5133,acmc-209dvn,4-ethylphenylsulfonyl chloride
570959
4-ethylbenzenesulfonyl chloride
CCC1=CC=C(C=C1)S(=O)(=O)Cl
Applications
4-Ethylbenzenesulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Reacts with water. Soluble in ethanol, acetone and toluene.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents, bases, amines and alcohols.
Melting Point 8°C to 12°C
Boiling Point 134°C to 136°C (7mmHg)
Quantity 100g
UN Number UN3265
Beilstein 2090130
Formula Weight 204.68
Percent Purity 97%
Chemical Name or Material 4-Ethylbenzenesulfonyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.