Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog Number p-7027708
Click to view available options
Quantity:
5 g
25 g
16712-69-9
C8H9ClO2S
204.668
MFCD00041480
LACFLXDRFOQEFZ-UHFFFAOYSA-N
4-ethylbenzene-1-sulfonyl chloride, p-ethylbenzenesulfonyl chloride, 4-ethylbenzenesulphonyl chloride, 4-ethyl-benzenesulfonyl chloride, ethylbenzenesulfonylchloride, 4-ethylbenzenesulfonylchloride, chloro 4-ethylphenyl sulfone, pubchem5133, acmc-209dvn, 4-ethylphenylsulfonyl chloride
570959
4-ethylbenzenesulfonyl chloride
CCC1=CC=C(C=C1)S(=O)(=O)Cl
16712-69-9
C8H9ClO2S
204.668
MFCD00041480
LACFLXDRFOQEFZ-UHFFFAOYSA-N
4-ethylbenzene-1-sulfonyl chloride, p-ethylbenzenesulfonyl chloride, 4-ethylbenzenesulphonyl chloride, 4-ethyl-benzenesulfonyl chloride, ethylbenzenesulfonylchloride, 4-ethylbenzenesulfonylchloride, chloro 4-ethylphenyl sulfone, pubchem5133, acmc-209dvn, 4-ethylphenylsulfonyl chloride
570959
4-ethylbenzenesulfonyl chloride
CCC1=CC=C(C=C1)S(=O)(=O)Cl

4-Ethylbenzenesulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethylbenzenesulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Reacts with water. Soluble in ethanol, acetone and toluene.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents, bases, amines and alcohols.
TRUSTED_SUSTAINABILITY
Melting Point 8°C to 12°C
Density 1.268
Boiling Point 134°C to 136°C (7 mmHg)
Refractive Index 1.545
Quantity 5 g
UN Number UN3265
Beilstein 2090130
Sensitivity Moisture sensitive
Solubility Information Reacts with water. Soluble in ethanol,acetone and toluene.
Formula Weight 204.68
Percent Purity 97%
Chemical Name or Material 4-Ethylbenzenesulfonyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit