Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethylbenzoic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
619-64-7
C9H10O2
150.18
MFCD00002570
ZQVKTHRQIXSMGY-UHFFFAOYSA-N
benzoic acid, 4-ethyl,p-ethylbenzoic acid,4-ethyl benzoic acid,4-ethyl-benzoic acid,unii-9tkm8p0g6r,9tkm8p0g6r,p-ethylbenzoate,4-ethyl benzoate,4-ethylbenzoicacid,p-ethylbenzoic acid;
12086
4-ethylbenzoic acid
CCC1=CC=C(C=C1)C(=O)O
619-64-7
C9H10O2
150.18
MFCD00002570
ZQVKTHRQIXSMGY-UHFFFAOYSA-N
benzoic acid, 4-ethyl,p-ethylbenzoic acid,4-ethyl benzoic acid,4-ethyl-benzoic acid,unii-9tkm8p0g6r,9tkm8p0g6r,p-ethylbenzoate,4-ethyl benzoate,4-ethylbenzoicacid,p-ethylbenzoic acid;
12086
4-ethylbenzoic acid
CCC1=CC=C(C=C1)C(=O)O
Color White to Beige
Infrared Spectrum Authentic
Quantity 25g
Melting Point 110.0°C to 114.0°C
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C2H5C6H4CO2H
Beilstein 09, 529
Solubility Information Solubility in water: slightly soluble
Formula Weight 150.18
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 4-Ethylbenzoic acid, 97%
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 210-605-3