Alfa Aesar™ 4-Ethylbenzophenone, 96%

Manufacturer: Alfa Aesar™ B2448418

Quantity 50g

 View more versions of this product

Catalog No. AAB2448418

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18220-90-1
    Molecular Formula C15H14O
    Formula Weight 210.28
    MDL Number MFCD00015177
    Beilstein 1949192
    Chemical Name or Material 4-Ethylbenzophenone

    Specifications

    Formula Weight 210.28
    Boiling Point 150°C (0.5mmHg)
    Molecular Formula C15H14O
    Quantity 50g
    Flash Point >110°C (230°F)
    MDL Number MFCD00015177
    CAS 18220-90-1
    Beilstein 1949192
    Chemical Name or Material 4-Ethylbenzophenone
    Assay 96%

    Description

    Applications
    It is used in pharmaceutical industry.

    Solubility
    Insoluble in water.

    Notes
    Store at room temperature. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit