Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Ethylbenzophenone, 96%

Manufacturer:  Alfa Aesar™ B2448409

Quantity 10g

 View more versions of this product

Catalog No. AAB2448409


  Check Availability
Add to cart

Specifications

Description

Applications
It is used in pharmaceutical industry.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

18220-90-1
210.28
1949192
C15H14O
MFCD00015177
4-Ethylbenzophenone

Specifications

18220-90-1
C15H14O
>110°C (230°F)
MFCD00015177
4-Ethylbenzophenone
150°C (0.5mmHg)
10g
210.28
1949192
96%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit