Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Ethylbenzyl chloride, 97%, stab. with calcium carbonate

Manufacturer:  Alfa Aesar™ H2790014

Quantity 25g

 View more versions of this product

Catalog No. AAH2790014


  Check Availability
Add to cart

Specifications

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

1467-05-6
154.64
UN3265
C9H11Cl
MFCD00039357
4-Ethylbenzyl chloride, stabilized with calcium carbonate

Specifications

1467-05-6
154.64
UN3265
25g
97%
C9H11Cl
MFCD00039357
111°C (26mmHg)
4-Ethylbenzyl chloride, stabilized with calcium carbonate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit