Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 4-Ethylbenzyl chloride, 97%, stab. with calcium carbonate
Click to view available options
Quantity:
5g
25g
Description
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Insoluble in water.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Insoluble in water.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
Specifications
Density | 1.04 |
Boiling Point | 111°C (26mmHg) |
Quantity | 5g |
UN Number | UN3265 |
Solubility Information | Insoluble in water. |
Formula Weight | 154.64 |
Percent Purity | 97% |
Chemical Name or Material | 4-Ethylbenzyl chloride, stabilized with calcium carbonate |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures