Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethylbenzylamine, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
7441-43-2
C9H13N
135.21
MFCD01529871
DGAGEFUEKIORSQ-UHFFFAOYSA-N
4-ethylbenzylamine,4-ethylphenyl methanamine,1-4-ethylphenyl methanamine,4-ethylphenyl methylamine,4-ethyl benzylamine,4-ethyl-benzylamine,4-ethyl-benzyl amine,benzenemethanamine, 4-ethyl
3870219
(4-ethylphenyl)methanamine
CCC1=CC=C(C=C1)CN
7441-43-2
C9H13N
135.21
MFCD01529871
DGAGEFUEKIORSQ-UHFFFAOYSA-N
4-ethylbenzylamine,4-ethylphenyl methanamine,1-4-ethylphenyl methanamine,4-ethylphenyl methylamine,4-ethyl benzylamine,4-ethyl-benzylamine,4-ethyl-benzyl amine,benzenemethanamine, 4-ethyl
3870219
(4-ethylphenyl)methanamine
CCC1=CC=C(C=C1)CN
Boiling Point 208°C (700.0mmHg)
Specific Gravity 0.883
Refractive Index 1.5280 to 1.53
Density 0.8830g/mL
Packaging Glass bottle
Quantity 5g
Color Colorless
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3CH2C6H4CH2NH2
Beilstein 12, IV, 2641
Solubility Information Solubility in water: soluble
Formula Weight 135.21
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 4-Ethylbenzylamine
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger