Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Ethylbiphenyl, 99%

Click to view available options
Quantity:
5g
5707-44-8
C14H14
182.266
MFCD00010236
SRQOBNUBCLPPPH-UHFFFAOYSA-N
4-ethylbiphenyl,4-ethyl-1,1'-biphenyl,p-ethylbiphenyl,1,1'-biphenyl, 4-ethyl,4-ethyl biphenyl,biphenyl, 4-ethyl,1,1'-biphenyl, 4-ethyl-9ci,pubchem2513,4-ethyl-1-phenylbenzene,acmc-1asq3
79786
1-ethyl-4-phenylbenzene
CCC1=CC=C(C=C1)C2=CC=CC=C2
5707-44-8
C14H14
182.266
MFCD00010236
SRQOBNUBCLPPPH-UHFFFAOYSA-N
4-ethylbiphenyl,4-ethyl-1,1'-biphenyl,p-ethylbiphenyl,1,1'-biphenyl, 4-ethyl,4-ethyl biphenyl,biphenyl, 4-ethyl,1,1'-biphenyl, 4-ethyl-9ci,pubchem2513,4-ethyl-1-phenylbenzene,acmc-1asq3
79786
1-ethyl-4-phenylbenzene
CCC1=CC=C(C=C1)C2=CC=CC=C2
Melting Point 34°C to 36°C
Boiling Point 140°C (15mmHg)
Quantity 5g
Beilstein 1904973
Formula Weight 182.27
Percent Purity 99%
Chemical Name or Material 4-Ethylbiphenyl