Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Ethyloctanoic acid, 97%

Click to view available options
Quantity:
2.5g
10g
16493-80-4
C10H20O2
172.268
MFCD00506494
PWKJMPFEQOHBAC-UHFFFAOYSA-N
octanoic acid, 4-ethyl,4-ethylcaprylic acid,4-ethyl octanoic acid,acmc-1bvq5,4-ethyloctanoicacid,ksc175o1l,4-ethyloctanoic acid, fg
61840
4-ethyloctanoic acid
CCCCC(CC)CCC(=O)O
16493-80-4
C10H20O2
172.268
MFCD00506494
PWKJMPFEQOHBAC-UHFFFAOYSA-N
octanoic acid, 4-ethyl,4-ethylcaprylic acid,4-ethyl octanoic acid,acmc-1bvq5,4-ethyloctanoicacid,ksc175o1l,4-ethyloctanoic acid, fg
61840
4-ethyloctanoic acid
CCCCC(CC)CCC(=O)O
Applications
Five sulfur-containing flavor compounds were synthesized by the reaction of 4-ethyloctanoyl chloride with sulfur-containing alcohols or mercaptans. 4-ethyloctanoic acid is identified from flue-cured Virginia tobacco. Mainly used as daily flavor,food flavor,industrial flavor, nutrient, stabilizers, surfactants and emulsifiers.

Solubility
Slightly soluble in water(0.13 mg/mL). Soluble in hexane

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Density 0.908
Quantity 2.5g
Refractive Index 1.439
Flash Point >100°C (212°F)
Boiling Point 142°C to 143°C (10mmHg)
Solubility Information Slightly soluble in water(0.13mg/mL). Soluble in hexane
Formula Weight 172.27
Percent Purity 97%
Chemical Name or Material 4-Ethyloctanoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 240-560-5

TSCA : Yes

Recommended Storage : Ambient temperatures