Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethylphenol, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
250g
1kg
5kg
Packaging:
Plastic bottle
Plastic drum
123-07-9
C8H10O
122.17
HXDOZKJGKXYMEW-UHFFFAOYSA-N
p-ethylphenol,phenol, 4-ethyl,1-ethyl-4-hydroxybenzene,phenol, p-ethyl,1-hydroxy-4-ethylbenzene,para-ethylphenol,4-hydroxyphenylethane,4-ethyl-phenol,4-hydroxyethylbenzene,paraethylphenol
31242
CHEBI:49584
4-ethylphenol
CCC1=CC=C(C=C1)O
123-07-9
C8H10O
122.17
HXDOZKJGKXYMEW-UHFFFAOYSA-N
p-ethylphenol,phenol, 4-ethyl,1-ethyl-4-hydroxybenzene,phenol, p-ethyl,1-hydroxy-4-ethylbenzene,para-ethylphenol,4-hydroxyphenylethane,4-ethyl-phenol,4-hydroxyethylbenzene,paraethylphenol
31242
CHEBI:49584
4-ethylphenol
CCC1=CC=C(C=C1)O
Color Brown to White
Boiling Point 218.0°C to 219.0°C
Melting Point 41.0°C to 46.0°C
Packaging Plastic bottle
Quantity 1kg
Infrared Spectrum Authentic
Flash Point 100°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C2H5C6H4OH
Beilstein 06,472
Solubility Information Solubility in water: 4900mg/l (25°C). Other solubilities: soluble in alcohol and ether
Formula Weight 122.17
Physical Form Crystalline Mass or Crystals
Percent Purity 97%
Chemical Name or Material 4-Ethylphenol
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger