Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Ethylphenyl isothiocyanate, 97%

Click to view available options
Quantity:
10g
18856-63-8
C9H9NS
163.238
MFCD00041101
XXLWCGRHJFEMQH-UHFFFAOYSA-N
4-ethylphenyl isothiocyanate,1-ethyl-4-isothiocyanato-benzene,benzene,1-ethyl-4-isothiocyanato,4-ethylbenzenisothiocyanate,acmc-20amit,2-p-isothio-cyanatophenyl ethane,#
140438
1-ethyl-4-isothiocyanatobenzene
CCC1=CC=C(C=C1)N=C=S
18856-63-8
C9H9NS
163.238
MFCD00041101
XXLWCGRHJFEMQH-UHFFFAOYSA-N
4-ethylphenyl isothiocyanate,1-ethyl-4-isothiocyanato-benzene,benzene,1-ethyl-4-isothiocyanato,4-ethylbenzenisothiocyanate,acmc-20amit,2-p-isothio-cyanatophenyl ethane,#
140438
1-ethyl-4-isothiocyanatobenzene
CCC1=CC=C(C=C1)N=C=S
Boiling Point 245°C
Odor Pungent
Flash Point >110°C (230°F)
Quantity 10g
UN Number UN2922
Beilstein 2802384
Formula Weight 163.24
Percent Purity 97%
Chemical Name or Material 4-Ethylphenyl isothiocyanate

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.