Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Ethylphenylacetylene, 99%

Manufacturer:  Alfa Aesar™ H2631314

Quantity 25g

 View more versions of this product

Catalog No. AAH2631314


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

40307-11-7
130.19
1-Ethyl-4-ethynylbenzene
C10H10
MFCD00173887
4-Ethylphenylacetylene

Specifications

71°C (160°F)
25g
C10H10
MFCD00173887
4-Ethylphenylacetylene
192°C
40307-11-7
130.19
1-Ethyl-4-ethynylbenzene
99%

Safety and Handling

SDS & Safety Documents

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit