Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethylphenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass Bottle
63139-21-9
C8H11BO2
149.98
RZCPLOMUUCFPQA-UHFFFAOYSA-N
4-ethylphenyl boronic acid, 4-ethylbenzeneboronic acid, 4-ethylphenyl boranediol, 4-ethylphenylboronicacid, p-ethylphenylboronic acid, boronic acid, 4-ethylphenyl, pubchem7884, 4-ethylbenzenboronic acid, acmc-1bik6
2734352
(4-ethylphenyl)boronic acid
B(C1=CC=C(C=C1)CC)(O)O
63139-21-9
C8H11BO2
149.98
RZCPLOMUUCFPQA-UHFFFAOYSA-N
4-ethylphenyl boronic acid, 4-ethylbenzeneboronic acid, 4-ethylphenyl boranediol, 4-ethylphenylboronicacid, p-ethylphenylboronic acid, boronic acid, 4-ethylphenyl, pubchem7884, 4-ethylbenzenboronic acid, acmc-1bik6
2734352
(4-ethylphenyl)boronic acid
B(C1=CC=C(C=C1)CC)(O)O
Infrared Spectrum Authentic
Packaging Glass Bottle
Color White
Quantity 1g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Solubility Information Solubility in water: .. Other solubilities: soluble in ether,thf,dmso,methanol
Formula Weight 149.98
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 4-Ethylphenylboronic acid
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning