Alfa Aesar™ 4-Ethylphenylmagnesium bromide, 0.5M in 2-MeTHF

Manufacturer: Alfa Aesar™ H54137-AE

 View more versions of this product

Catalog No. AAH54137AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22873-28-5
    Molecular Formula C8H9BrMg
    Formula Weight 209.36g/mol
    MDL Number MFCD01311467
    UN Number UN2924
    Chemical Name or Material 4-Ethylphenylmagnesium bromide

    Specifications

    Concentration 0.5M in 2-MeTHF
    Quantity 100mL
    CAS 22873-28-5
    Molecular Formula C8H9BrMg
    Formula Weight 209.36g/mol
    MDL Number MFCD01311467
    UN Number UN2924
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H314
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Chemical Name or Material 4-Ethylphenylmagnesium bromide

    Description

    Applications
    4-Ethylphenylmagnesium bromide, 0.5M in 2-MeTHF is used as a pharmaceutical intermediate.

    Solubility
    Reacts with water.

    Notes
    Air and moisture sensitive. Store under inert gas. Store away from oxidizing agents, air, water/ moisture, bases, heat.

    GHS H Statement
    H225-H314
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.

    2° to 8°C

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit