Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Acros Organics

Click to view available options
Quantity:
50mL
Packaging:
AcroSeal™ Glass Bottle
22873-28-5,109-99-9
C8H9BrMg
209.37
QSGSHZBZGXYVOQ-UHFFFAOYSA-M
4-ethylphenylmagnesium bromide,4-ethylphenyl magnesium bromide,vlouakpiqasfds-uhfffaoysa-m,4-ethylphenylmagnesium bromide, 0.5m in 2-methf,4-ethylphenylmagnesium bromide 0.5 m in tetrahydrofuran,4-ethylphenylmagnesium bromide solution, 0.5 m in thf
11241259
magnesium;ethylbenzene;bromide
CCC1=CC=[C-]C=C1.[Mg+2].[Br-]
22873-28-5,109-99-9
C8H9BrMg
209.37
QSGSHZBZGXYVOQ-UHFFFAOYSA-M
4-ethylphenylmagnesium bromide,4-ethylphenyl magnesium bromide,vlouakpiqasfds-uhfffaoysa-m,4-ethylphenylmagnesium bromide, 0.5m in 2-methf,4-ethylphenylmagnesium bromide 0.5 m in tetrahydrofuran,4-ethylphenylmagnesium bromide solution, 0.5 m in thf
11241259
magnesium;ethylbenzene;bromide
CCC1=CC=[C-]C=C1.[Mg+2].[Br-]
Quantity 50mL
Color Yellow to Brown
Density 0.9500g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.95
CAS Min % 8.82,86.78
CAS Max % 13.22,91.18
Linear Formula CH3CH2C6H4MgBr
Solubility Information Solubility in water: reacts
Formula Weight 209.37
Physical Form Solution
Chemical Name or Material 4-Ethylphenylmagnesium bromide
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger