Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethylpyridine 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
536-75-4
C7H9N
107.15
MFCD00006450
VJXRKZJMGVSXPX-UHFFFAOYSA-N
pyridine, 4-ethyl,.gamma.-ethylpyridine,4-ethyl pyridine,unii-3kw0se2tl5,4-ethyl-pyridine,pyridine, c1-3-alkyl derivs.,3kw0se2tl5,gamma-ethylpyridine,c1-c3 alkylpyridine,4ethylpyridine
10822
4-ethylpyridine
CCC1=CC=NC=C1
536-75-4
C7H9N
107.15
MFCD00006450
VJXRKZJMGVSXPX-UHFFFAOYSA-N
pyridine, 4-ethyl,.gamma.-ethylpyridine,4-ethyl pyridine,unii-3kw0se2tl5,4-ethyl-pyridine,pyridine, c1-3-alkyl derivs.,3kw0se2tl5,gamma-ethylpyridine,c1-c3 alkylpyridine,4ethylpyridine
10822
4-ethylpyridine
CCC1=CC=NC=C1
Melting Point -91°C
Boiling Point 164°C to 170°C
Flash Point 47°C
Specific Gravity 0.942
Refractive Index 1.5009 to 1.5029
Density 0.9420g/mL
Color Colorless to Yellow
Quantity 100g
Packaging Glass bottle
Infrared Spectrum Authentic
Water 0.5% max. (K.F.)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (C5H3N)CH2CH3
Beilstein 20, 243
Merck Index 15, 3901
Solubility Information Solubility in water: ca 34g/L in water. Other solubilities: soluble in alcohol and ether
Formula Weight 107.15
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 4-Ethylpyridine, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-646-7

TSCA : TSCA