Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethylresorcinol 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2896-60-8
C8H10O2
138.17
MFCD00002283
VGMJYYDKPUPTID-UHFFFAOYSA-N
4-ethylresorcinol,1,3-benzenediol, 4-ethyl,6-ethylresorcinol,resorcinol, 4-ethyl,1,3-dihydroxy-4-ethylbenzene,4-ethyl resorcinol,2,4-dihydroxy-1-ethylbenzene,unii-8ye81t06d5,4-ethyl-benzene-1,3-diol,4-ethyl-resorcinol
17927
4-ethylbenzene-1,3-diol
CCC1=C(C=C(C=C1)O)O
2896-60-8
C8H10O2
138.17
MFCD00002283
VGMJYYDKPUPTID-UHFFFAOYSA-N
4-ethylresorcinol,1,3-benzenediol, 4-ethyl,6-ethylresorcinol,resorcinol, 4-ethyl,1,3-dihydroxy-4-ethylbenzene,4-ethyl resorcinol,2,4-dihydroxy-1-ethylbenzene,unii-8ye81t06d5,4-ethyl-benzene-1,3-diol,4-ethyl-resorcinol
17927
4-ethylbenzene-1,3-diol
CCC1=C(C=C(C=C1)O)O
Quantity 25g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Beige
Melting Point 95.0°C to 99.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 06, I, 441
Formula Weight 138.17
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Ethylresorcinol, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 220-777-1

TSCA : TSCA