Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4'-Ethynylacetophenone, 98%, Acros Organics
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
Specifications
Color | Yellow |
Packaging | Glass bottle |
Quantity | 1g |
Melting Point | 67.0°C to 70.0°C |
Infrared Spectrum | Authentic |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97.5% min. (GC) |
Linear Formula | CHCC6H4COCH3 |
Formula Weight | 144.17 |
Show More |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If eye irritation persists: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective glov
GHS Signal Word: Warning