Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Ethynylacetophenone, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
42472-69-5
C10H8O
144.17
MFCD04974025
WHFPFLKZXFBCSO-UHFFFAOYSA-N
1-4-ethynylphenyl ethanone,1-4-ethynyl-phenyl-ethanone,4-acetylphenylacetylene,ethanone, 1-4-ethynylphenyl,4'-ethynylacetophenone,1-4-ethynylphenyl ethan-1-one,4-acetylphenylethyne,4-acetylphenyl acetylene,1-4-ethynylphenyl-ethanone,ethanone,1-4-ethynylphenyl
5123626
1-(4-ethynylphenyl)ethanone
CC(=O)C1=CC=C(C=C1)C#C
42472-69-5
C10H8O
144.17
MFCD04974025
WHFPFLKZXFBCSO-UHFFFAOYSA-N
1-4-ethynylphenyl ethanone,1-4-ethynyl-phenyl-ethanone,4-acetylphenylacetylene,ethanone, 1-4-ethynylphenyl,4'-ethynylacetophenone,1-4-ethynylphenyl ethan-1-one,4-acetylphenylethyne,4-acetylphenyl acetylene,1-4-ethynylphenyl-ethanone,ethanone,1-4-ethynylphenyl
5123626
1-(4-ethynylphenyl)ethanone
CC(=O)C1=CC=C(C=C1)C#C
Infrared Spectrum Authentic
Quantity 5g
Color Yellow
Melting Point 67.0°C to 70.0°C
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CHCC6H4COCH3
Formula Weight 144.17
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 4'-Ethynylacetophenone
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If eye irritation persists: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective glov

GHS Signal Word: Warning