Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethynylanisole, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
768-60-5
C9H8O
132.16
MFCD00168815
KBIAVTUACPKPFJ-UHFFFAOYSA-N
4-ethynylanisole,4-methoxyphenylacetylene,4'-methoxyphenyl acetylene,1-ethynyl-4-methoxy-benzene,1-eth-1-ynyl-4-methoxybenzene,benzene, 1-ethynyl-4-methoxy,4-methoxyphenyl acetylene,4-ethynyl-1-methoxybenzene,4'-methoxyphenylacetylene,p-ethynylanisole
251020
1-ethynyl-4-methoxybenzene
COC1=CC=C(C=C1)C#C
768-60-5
C9H8O
132.16
MFCD00168815
KBIAVTUACPKPFJ-UHFFFAOYSA-N
4-ethynylanisole,4-methoxyphenylacetylene,4'-methoxyphenyl acetylene,1-ethynyl-4-methoxy-benzene,1-eth-1-ynyl-4-methoxybenzene,benzene, 1-ethynyl-4-methoxy,4-methoxyphenyl acetylene,4-ethynyl-1-methoxybenzene,4'-methoxyphenylacetylene,p-ethynylanisole
251020
1-ethynyl-4-methoxybenzene
COC1=CC=C(C=C1)C#C
Melting Point 28°C to 30°C
Flash Point 82°C
Boiling Point 87°C to 91°C (11.0mmHg)
Specific Gravity 1.019
Density 1.0190g/mL
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Brown or Colorless to Yellow
Refractive Index 1.5625 to 1.5675 (20°C, 589nm)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula HC∽C6H4OCH3
Solubility Information Solubility in water: insoluble
Formula Weight 132.16
Physical Form Liquid or Low Melting Solid
Percent Purity 97%
Chemical Name or Material 4-Ethynylanisole
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.