Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethynylbenzeneboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog Number p-7034096
Click to view available options
Quantity:
1 g
5 g
1034287-04-1
C14H17BO2
228.098
MFCD16294504
LOVNTFMVZVIASV-UHFFFAOYSA-N
2-4-ethynylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-ethynyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 4-ethynylbenzeneboronic acid pinacol ester, amtb205, 4-ethynylphenylboronic acid pinacol ester
57415690
2-(4-ethynylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C#C
1034287-04-1
C14H17BO2
228.098
MFCD16294504
LOVNTFMVZVIASV-UHFFFAOYSA-N
2-4-ethynylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-ethynyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 4-ethynylbenzeneboronic acid pinacol ester, amtb205, 4-ethynylphenylboronic acid pinacol ester
57415690
2-(4-ethynylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C#C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 228.1
Percent Purity 95%
Chemical Name or Material 4-Ethynylbenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit